Assay Details
| Gene Symbol: CTDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039674.1 | XP_017895163.1 | 4 - 5 | 772 | 52 | |
| XM_018039675.1 | XP_017895164.1 | 4 - 5 | 772 | 52 | ||
| XM_018039676.1 | XP_017895165.1 | 4 - 5 | 484 | 52 | ||
More Information
Set Membership: |
|

CTDP1
Genomic Map