Assay Details
| Gene Symbol: DECR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040919.1 | XP_017896408.1 | 4 - 5 | 480 | 57 | |
| GenBank mRNA | JO427022.1 | - | 4 - 5 | 573 | 57 | |
| JO599827.2 | - | 2 - 3 | 217 | 57 | ||
More Information
Set Membership: |
|

DECR2
Genomic Map