Assay Details
| Gene Symbol: IRF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043034.1 | XP_017898523.1 | 6 - 7 | 1029 | 70 | |
| GenBank mRNA | JO632476.2 | - | 6 - 7 | 1003 | 70 | |
More Information
Set Membership: |
|

IRF7
Genomic Map