Assay Details
| Gene Symbol: A4GNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675522.3 | XP_005675579.1 | 2 - 3 | 953 | 67 | |
| XM_018049971.1 | XP_017905460.1 | 2 - 3 | 710 | 67 | ||
More Information
Set Membership: |
|

A4GNT
Genomic Map