Assay Details
| Gene Symbol: FAM168B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065964.1 | XP_017921453.1 | 3 - 4 | 311 | 88 | |
| GenBank mRNA | JO438073.2 | - | 3 - 4 | 444 | 88 | |
| JO643310.2 | - | 3 - 4 | 399 | 88 | ||
More Information
Set Membership: |
|

FAM168B
Genomic Map