Assay Details
| Gene Symbol: C5H12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680229.2 | XP_005680286.2 | 14 - 15 | 1783 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5H12orf40| Gene Symbol: C5H12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680229.2 | XP_005680286.2 | 14 - 15 | 1783 | 117 | |
Set Membership: |
|