Assay Details
| Gene Symbol: KCNMB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680231.3 | XP_005680288.2 | 2 - 3 | 793 | 74 | |
| GenBank mRNA | JO628296.2 | - | 2 - 3 | 495 | 74 | |
| JT733029.1 | - | 5 - 6 | 936 | 74 | ||
More Information
Set Membership: |
|

KCNMB4
Genomic Map