Assay Details
| Gene Symbol: LOC102175297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685071.3 | XP_005685128.1 | 2 - 3 | 384 | 72 | |
| GenBank mRNA | JO419757.2 | - | 6 - 7 | 1614 | 72 | |
More Information
Set Membership: |
|

LOC102175297
Genomic Map