Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685736.3 | XP_005685793.1 | 5 - 6 | 945 | 65 | |
| XM_005685737.3 | XP_005685794.1 | 5 - 6 | 890 | 65 | ||
| XM_005685738.3 | XP_005685795.1 | 5 - 6 | 1196 | 65 | ||
More Information
Set Membership: |
|

DNAAF4
Genomic Map