Assay Details
| Gene Symbol: TP53I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686955.3 | XP_005687012.1 | 2 - 3 | 841 | 71 | |
| XM_005686956.3 | XP_005687013.1 | 3 - 4 | 542 | 71 | ||
More Information
Set Membership: |
|

TP53I3
Genomic Map