Assay Details
| Gene Symbol: IAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055679.1 | XP_017911168.1 | 5 - 6 | 619 | 74 | |
| XM_018055680.1 | XP_017911169.1 | 5 - 6 | 633 | 74 | ||
| GenBank mRNA | JO355053.2 | - | 1 - 2 | 354 | 74 | |
| JO515710.2 | - | 3 - 4 | 326 | 74 | ||
More Information
Set Membership: |
|

IAH1
Genomic Map