Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689233.3 | XP_005689290.1 | 10 - 11 | 2745 | 89 | |
| XM_005689234.3 | XP_005689291.1 | 10 - 11 | 2745 | 89 | ||
| XM_018058232.1 | XP_017913721.1 | 6 - 7 | 2213 | 89 | ||
| GenBank mRNA | JT707671.1 | - | 10 - 11 | 2748 | 89 | |
More Information
Set Membership: |
|

VIRMA
Genomic Map