Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689233.3 | XP_005689290.1 | 19 - 20 | 4619 | 106 | |
| XM_005689234.3 | XP_005689291.1 | 18 - 19 | 4466 | 106 | ||
| XM_018058232.1 | XP_017913721.1 | 15 - 16 | 4087 | 106 | ||
More Information
Set Membership: |
|

VIRMA
Genomic Map