Assay Details
| Gene Symbol: LOC102183983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689293.2 | XP_005689350.2 | 1 - 2 | 80 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102183983| Gene Symbol: LOC102183983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689293.2 | XP_005689350.2 | 1 - 2 | 80 | 101 | |
Set Membership: |
|