Assay Details
Gene Symbol: SLC30A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689295.2 | XP_005689352.2 | 3 - 4 | 440 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
