Assay Details
| Gene Symbol: LOC102173866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690001.2 | XP_005690058.1 | 3 - 4 | 531 | 107 | |
| GenBank mRNA | KA306658.1 | - | 1 - 2 | 148 | 107 | |
More Information
Set Membership: |
|

LOC102173866
Genomic Map