Assay Details
| Gene Symbol: CA5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691856.3 | XP_005691913.1 | 3 - 4 | 557 | 67 | |
| GenBank mRNA | JO402918.2 | - | 5 - 6 | 563 | 67 | |
| JO402919.2 | - | 4 - 5 | 575 | 67 | ||
More Information
Set Membership: |
|

CA5A
Genomic Map