Assay Details
| Gene Symbol: ADGRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063154.1 | XP_017918643.1 | 2 - 3 | 212 | 77 | |
| GenBank mRNA | JO325199.2 | - | 7 - 8 | 700 | 77 | |
More Information
Set Membership: |
|

ADGRG3
Genomic Map