Assay Details
| Gene Symbol: SMIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063806.1 | XP_017919295.1 | 2 - 3 | 3113 | 66 | |
| XM_018063807.1 | XP_017919296.1 | 2 - 3 | 317 | 66 | ||
| XM_018063808.1 | XP_017919297.1 | 2 - 3 | 1166 | 66 | ||
| GenBank mRNA | JO639208.2 | - | 1 - 2 | 784 | 66 | |
More Information
Set Membership: |
|

SMIM5
Genomic Map