Assay Details
| Gene Symbol: MAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040367.1 | XP_017895856.1 | 3 - 4 | 1278 | 69 | |
| XM_018040368.1 | XP_017895857.1 | 3 - 4 | 1177 | 69 | ||
| XM_018040370.1 | XP_017895859.1 | 3 - 4 | 1282 | 69 | ||
| XM_018040371.1 | XP_017895860.1 | 3 - 4 | 1177 | 69 | ||
| XM_018040372.1 | XP_017895861.1 | 4 - 5 | 1117 | 69 | ||
| XM_018040373.1 | XP_017895862.1 | 4 - 5 | 1117 | 69 | ||
| GenBank mRNA | KA347009.1 | - | 3 - 4 | 1024 | 69 | |
More Information
Set Membership: |
|

MAZ
Genomic Map