Assay Details
Gene Symbol: SHTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013975257.2 | XP_013830711.2 | 12 - 13 | 1244 | 62 | |
XM_018041468.1 | XP_017896957.1 | 12 - 13 | 1244 | 62 | ||
XM_018041469.1 | XP_017896958.1 | 12 - 13 | 1229 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
