Assay Details
| Gene Symbol: ZP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699001.2 | XP_005699058.1 | 7 - 8 | 968 | 64 | |
| XM_005699002.3 | XP_005699059.1 | 7 - 8 | 973 | 64 | ||
More Information
Set Membership: |
|

ZP4
Genomic Map