Assay Details
| Gene Symbol: ZNF280D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054248.1 | XP_017909737.1 | 5 - 6 | 535 | 89 | |
| XM_018054249.1 | XP_017909738.1 | 4 - 5 | 557 | 89 | ||
| XM_018054250.1 | XP_017909739.1 | 5 - 6 | 409 | 89 | ||
| XM_018054251.1 | XP_017909740.1 | 4 - 5 | 489 | 89 | ||
| XM_018054252.1 | XP_017909741.1 | 3 - 4 | 337 | 89 | ||
| XM_018054253.1 | XP_017909742.1 | 5 - 6 | 535 | 89 | ||
| XM_018054255.1 | XP_017909744.1 | 5 - 6 | 465 | 89 | ||
| XM_018054256.1 | XP_017909745.1 | 5 - 6 | 465 | 89 | ||
| GenBank mRNA | JO426236.1 | - | 17 - 18 | 2927 | 89 | |
More Information
Set Membership: |
|

ZNF280D
Genomic Map