Assay Details
| Gene Symbol: DNAAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054378.1 | XP_017909867.1 | 2 - 3 | 4387 | 103 | |
| GenBank mRNA | JO437708.2 | - | 2 - 3 | 822 | 103 | |
| JO592565.2 | - | 1 - 2 | 526 | 103 | ||
More Information
Set Membership: |
|

DNAAF2
Genomic Map