Assay Details
| Gene Symbol: AHCY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057737.1 | XP_017913226.1 | 5 - 6 | 659 | 59 | |
| GenBank mRNA | JO580773.1 | - | 5 - 6 | 1547 | 59 | |
| JO645767.2 | - | 5 - 6 | 1488 | 59 | ||
More Information
Set Membership: |
|

AHCY
Genomic Map