Assay Details
| Gene Symbol: CD55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060124.1 | XP_017915613.1 | 6 - 7 | 1193 | 59 | |
| XM_018060125.1 | XP_017915614.1 | 6 - 7 | 1194 | 59 | ||
| XM_018060126.1 | XP_017915615.1 | 6 - 7 | 1194 | 59 | ||
| XM_018060127.1 | XP_017915616.1 | 6 - 7 | 1192 | 59 | ||
| XM_018060128.1 | XP_017915617.1 | 6 - 7 | 1192 | 59 | ||
| XM_018060129.1 | XP_017915618.1 | 6 - 7 | 1192 | 59 | ||
| XM_018060130.1 | XP_017915619.1 | 6 - 7 | 1192 | 59 | ||
| XM_018060131.1 | XP_017915620.1 | 6 - 7 | 1192 | 59 | ||
| XM_018060132.1 | XP_017915621.1 | 6 - 7 | 1189 | 59 | ||
| XM_018060134.1 | XP_017915623.1 | 6 - 7 | 1190 | 59 | ||
| XM_018060135.1 | XP_017915624.1 | 6 - 7 | 1191 | 59 | ||
| XM_018060136.1 | XP_017915625.1 | 6 - 7 | 1189 | 59 | ||
More Information
Set Membership: |
|

CD55
Genomic Map