Assay Details
| Gene Symbol: HSPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063084.1 | XP_017918573.1 | 3 - 4 | 653 | 69 | |
| XM_018063085.1 | XP_017918574.1 | 3 - 4 | 656 | 69 | ||
| GenBank mRNA | JO637032.2 | - | 5 - 6 | 1067 | 69 | |
More Information
Set Membership: |
|

HSPBP1
Genomic Map