Assay Details
| Gene Symbol: ADAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971931.2 | XP_013827385.2 | 10 - 11 | 1160 | 64 | |
| XM_018064258.1 | XP_017919747.1 | 10 - 11 | 1292 | 64 | ||
| XM_018064259.1 | XP_017919748.1 | 10 - 11 | 1223 | 64 | ||
| XM_018064260.1 | XP_017919749.1 | 9 - 10 | 1145 | 64 | ||
| XM_018064261.1 | XP_017919750.1 | 9 - 10 | 1044 | 64 | ||
| GenBank mRNA | JO418590.2 | - | 1 - 2 | 936 | 64 | |
More Information
Set Membership: |
|

ADAP2
Genomic Map