Assay Details
| Gene Symbol: RCAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055819.1 | XP_017911308.1 | 2 - 3 | 558 | 90 | |
| XM_018055891.1 | XP_017911380.1 | 2 - 3 | 498 | 90 | ||
| GenBank mRNA | JO363489.2 | - | 2 - 3 | 362 | 90 | |
| JO363490.2 | - | 2 - 3 | 362 | 90 | ||
| JO363491.2 | - | 2 - 3 | 362 | 90 | ||
| JO380523.2 | - | 2 - 3 | 452 | 90 | ||
| JO380527.2 | - | 2 - 3 | 452 | 90 | ||
| JO412632.2 | - | 2 - 3 | 325 | 90 | ||
| JO412633.2 | - | 2 - 3 | 426 | 90 | ||
| JO451505.2 | - | 2 - 3 | 566 | 90 | ||
| JO536800.2 | - | 2 - 3 | 369 | 90 | ||
| JO637564.2 | - | 2 - 3 | 611 | 90 | ||
| JO637565.2 | - | 2 - 3 | 611 | 90 | ||
| JT705616.1 | - | 1 - 2 | 116 | 90 | ||
| JT716551.1 | - | 3 - 4 | 608 | 90 | ||
| JT738539.1 | - | 2 - 3 | 331 | 90 | ||
| KA313262.1 | - | 2 - 3 | 891 | 90 | ||
More Information
Set Membership: |
|

RCAN1
Genomic Map