Assay Details
| Gene Symbol: NES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046299.1 | XP_017901788.1 | 3 - 4 | 1111 | 66 | |
| GenBank mRNA | JO430480.2 | - | 1 - 2 | 1316 | 66 | |
More Information
Set Membership: |
|

NES
Genomic Map