Assay Details
| Gene Symbol: S100B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675677.3 | XP_005675734.1 | 2 - 3 | 245 | 85 | |
| GenBank mRNA | JT763336.1 | - | 1 - 2 | 1296 | 85 | |
| JT763337.1 | - | 1 - 2 | 1295 | 85 | ||
| KC244280.1 | - | 1 - 2 | 141 | 85 | ||
More Information
Set Membership: |
|

S100B
Genomic Map