Assay Details
| Gene Symbol: AUNIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676829.2 | XP_005676886.2 | 2 - 3 | 315 | 58 | |
| GenBank mRNA | KA344980.1 | - | 1 - 2 | 503 | 58 | |
More Information
Set Membership: |
|

AUNIP
Genomic Map