Assay Details
| Gene Symbol: TSFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048002.1 | XP_017903491.1 | 3 - 4 | 454 | 91 | |
| GenBank mRNA | JO437927.2 | - | 3 - 4 | 469 | 91 | |
| JO629111.2 | - | 3 - 4 | 756 | 91 | ||
More Information
Set Membership: |
|

TSFM
Genomic Map