Assay Details
| Gene Symbol: NR1H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680501.3 | XP_005680558.1 | 8 - 9 | 2566 | 73 | |
| XM_005680502.3 | XP_005680559.1 | 8 - 9 | 2555 | 73 | ||
| XM_018048151.1 | XP_017903640.1 | 10 - 11 | 1480 | 73 | ||
| XM_018048152.1 | XP_017903641.1 | 10 - 11 | 1468 | 73 | ||
| GenBank mRNA | JO421456.2 | - | 1 - 2 | 525 | 73 | |
| JT666907.1 | - | 9 - 10 | 1368 | 73 | ||
More Information
Set Membership: |
|

NR1H4
Genomic Map