Assay Details
| Gene Symbol: RDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686006.2 | XP_005686063.2 | 5 - 6 | 845 | 51 | |
| GenBank mRNA | KA318115.1 | - | 5 - 6 | 828 | 51 | |
More Information
Set Membership: |
|

RDH12
Genomic Map