Assay Details
| Gene Symbol: CIPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053926.1 | XP_017909415.1 | 3 - 4 | 544 | 63 | |
| GenBank mRNA | JO631008.2 | - | 3 - 4 | 520 | 63 | |
More Information
Set Membership: |
|

CIPC
Genomic Map