Assay Details
| Gene Symbol: BEND7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057122.1 | XP_017912611.1 | 4 - 5 | 998 | 78 | |
| XM_018057123.1 | XP_017912612.1 | 4 - 5 | 998 | 78 | ||
| XM_018057124.1 | XP_017912613.1 | 3 - 4 | 699 | 78 | ||
| XM_018057125.1 | XP_017912614.1 | 4 - 5 | 998 | 78 | ||
| XM_018057126.1 | XP_017912615.1 | 4 - 5 | 1003 | 78 | ||
| GenBank mRNA | JO428481.1 | - | 3 - 4 | 668 | 78 | |
More Information
Set Membership: |
|

BEND7
Genomic Map