Assay Details
| Gene Symbol: HAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688179.3 | XP_005688236.1 | 4 - 5 | 749 | 66 | |
| XM_005688180.3 | XP_005688237.2 | 3 - 4 | 500 | 66 | ||
| GenBank mRNA | JO426350.1 | - | 4 - 5 | 2463 | 66 | |
More Information
Set Membership: |
|

HAO1
Genomic Map