Assay Details
| Gene Symbol: ZHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058531.1 | XP_017914020.1 | 3 - 4 | 4857 | 117 | |
| XM_018058532.1 | XP_017914021.1 | 4 - 5 | 5013 | 117 | ||
More Information
Set Membership: |
|

ZHX2
Genomic Map