Assay Details
| Gene Symbol: ATP6V1G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691060.3 | XP_005691117.1 | 2 - 3 | 306 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP6V1G3| Gene Symbol: ATP6V1G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691060.3 | XP_005691117.1 | 2 - 3 | 306 | 88 | |
Set Membership: |
|