Assay Details
| Gene Symbol: TXNL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692203.3 | XP_005692260.1 | 3 - 4 | 567 | 84 | |
| XM_018062222.1 | XP_017917711.1 | 3 - 4 | 393 | 84 | ||
| GenBank mRNA | JO601951.2 | - | 1 - 2 | 778 | 84 | |
| KA308423.1 | - | 3 - 4 | 502 | 84 | ||
More Information
Set Membership: |
|

TXNL4B
Genomic Map