Assay Details
| Gene Symbol: UBE2G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064440.1 | XP_017919929.1 | 4 - 5 | 767 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2G1| Gene Symbol: UBE2G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064440.1 | XP_017919929.1 | 4 - 5 | 767 | 82 | |
Set Membership: |
|