Assay Details
| Gene Symbol: ISG20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694942.3 | XP_005694999.1 | 2 - 3 | 518 | 68 | |
| GenBank mRNA | JO318540.2 | - | 2 - 3 | 382 | 68 | |
| JO599637.2 | - | 2 - 3 | 399 | 68 | ||
More Information
Set Membership: |
|

ISG20
Genomic Map