Assay Details
| Gene Symbol: STX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041038.1 | XP_017896527.1 | 7 - 8 | 597 | 56 | |
| XM_018041039.1 | XP_017896528.1 | 7 - 8 | 590 | 56 | ||
| GenBank mRNA | JO626976.2 | - | 3 - 4 | 438 | 56 | |
More Information
Set Membership: |
|

STX1A
Genomic Map