Assay Details
| Gene Symbol: UPK3BL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697803.3 | XP_005697860.3 | 3 - 4 | 843 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UPK3BL2| Gene Symbol: UPK3BL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697803.3 | XP_005697860.3 | 3 - 4 | 843 | 82 | |
Set Membership: |
|