Assay Details
| Gene Symbol: PANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698201.3 | XP_005698258.1 | 4 - 5 | 986 | 72 | |
| XM_018041474.1 | XP_017896963.1 | 4 - 5 | 1631 | 72 | ||
| XM_018041475.1 | XP_017896964.1 | 6 - 7 | 1392 | 72 | ||
| XM_018041476.1 | XP_017896965.1 | 6 - 7 | 1243 | 72 | ||
| GenBank mRNA | JO601071.2 | - | 6 - 7 | 1197 | 72 | |
More Information
Set Membership: |
|

PANK1
Genomic Map