Assay Details
| Gene Symbol: OVOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043496.1 | XP_017898985.1 | 2 - 3 | 670 | 74 | |
| GenBank mRNA | JI861834.1 | - | 2 - 3 | 320 | 74 | |
| JT754443.1 | - | 2 - 3 | 328 | 74 | ||
More Information
Set Membership: |
|

OVOL1
Genomic Map