Assay Details
| Gene Symbol: OTUD6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700710.1 | XP_005700767.1 | 1 | 621 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTUD6A| Gene Symbol: OTUD6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700710.1 | XP_005700767.1 | 1 | 621 | 56 | |
Set Membership: |
|