Assay Details
| Gene Symbol: SEPT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048829.1 | XP_017904318.1 | 3 - 4 | 466 | 64 | |
| XM_018048830.1 | XP_017904319.1 | 3 - 4 | 480 | 64 | ||
| XM_018048831.1 | XP_017904320.1 | 4 - 5 | 606 | 64 | ||
| GenBank mRNA | JO632795.2 | - | 3 - 4 | 595 | 64 | |
| JO632796.2 | - | 4 - 5 | 722 | 64 | ||
More Information
Set Membership: |
|

SEPT3
Genomic Map