Assay Details
| Gene Symbol: BST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049746.1 | XP_017905235.1 | 3 - 4 | 689 | 66 | |
| XM_018049747.1 | XP_017905236.1 | 3 - 4 | 687 | 66 | ||
| GenBank mRNA | JO603279.2 | - | 3 - 4 | 601 | 66 | |
More Information
Set Membership: |
|

BST1
Genomic Map